Lyijy 1000 mg/l laimeassa typpihapossa (from Pb) standard for AAS
Toimittaja: Avantor
BAKR6930.0100EA 126 EUR
BAKR6930.0100
Lyijy 1000 mg/l laimeassa typpihapossa (from Pb) standard for AAS
Sertifikaatit: Traceable to NIST.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022