Procaine hydrochloride

Toimittaja: EDQM

EDQMP3100000EA 251 EUR
EDQMP3100000
Procaine hydrochloride
Procaine hydrochloride
Kaava: C₁₃H₂₁ClN₂O₂
MDL numero: MFCD00013000
CAS-numero: 51-05-8
EINECS: 200-077-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR