VWR® Traceable®, Software
Toimittaja: VWR
VWR® Traceable®
630-2507EA 606 EUR
630-2507
VWR® Traceable®, Software
Ohjelmisto Laboratorio-ohjelmisto
Data capture program for thermometers communication port. The software allows connection of up to eight instruments all running simultaneously on one computer. Records interval readings from 1 to 10 000 seconds; displays minimum/maximum readings; uses an alarm mode that allows the user to be notified visually, audibly and by email when an alarm is triggered. Readings are saved to a file that can be viewed and printed as is, or may be printed in any report or spreadsheet format. The software is designed to work with Windows® 98/Me/NT/2000/XP/Vista/Windows 7.
  • 24/7 notification of audible/visual alarms are sent to user’s computer, remote computer, or via email anywhere
  • Simply connect the cable to the instrument and USB port (or serial port), and data is captured
  • Captures data in the background which enables the user to work on other programs
Toimitustiedot: Includes a CD, a 1,8 m cable (USB, serial, and instrument connections), cable plugs into the instrument and computer.