Assortment of remainder tubing, BOLA
Toimittaja: Bohlender
BOHLS1899-10EA 224 EUR
BOHLS1899-10
Assortment of remainder tubing, BOLA
PTFE, FEP, PFA.
  • Very good chemical resistance
  • Very good temperature resistance
No rejections. Approx. 10 rolls of unsorted tubing made of PTFE, FEP and PFA with different diameters. Length of up to 2 metres per roll. No selection possible.