DNA transfection reagent, jetPEI®-Macrophage
Toimittaja: Polyplus-transfection
jetPEI®
101000043.EA 0 EUR
101000043.
DNA transfection reagent, jetPEI®-Macrophage
Transfektioreagenssit
jetPEI®-Macrophage DNA transfection reagent is dedicated to the transfection of macrophage primary cells and cell lines which express mannose receptors, such as RAW 264.7.
  • Efficient transfection of primary macrophages, glial and dendritic cells
  • Easy-to-use protocol
  • Compatible with serum and antibiotics
Since 1st of September 2021 Polyplus-Tranfection changed product part numbers. Products are remaining the same and the change is only related to the product part number. New product part numbers have symbol “New”. For duplicated products with symbol “telephone” please be informed that it’s the same product but with previous part number which we will only offer till the end of stock.
Sertifikaatit: Polyplus-transfection® has been an ISO 9001 accredited company since 2002. Since then this certification has been successfully renewed every three years.
Every batch of jetPEI®-Macrophage is tested in a transfection assay. Typically, transfection of a firefly luciferase gene under the control of the CMV promote gives 109 RLU (relative light unit)/mg of protein. The value for each batch is indicated on the Certificate of Analysis.
Toimitustiedot: 150 mM NaCl solution is included. 0,5 ml of jetPEI®-Macrophage transfection reagent is sufficient to perform up to 250 transfections in 24-well plates.