Hait: Puhdastilamatot
Koko lattiapinnalle asennettavia pysyviä puhdastilamattoja.
Koko lattiapinnalle asennettavia pysyviä puhdastilamattoja.
17  results were found
SearchResultCount:"17"
Sort Results
Luettelonäkymä Easy View (new)
Rate These Search Results
Toimittaja: VWR Collection
Kuvaus: Permanent mats are installed to provide the highest level of cleanliness. The life of mats strongly depends on the volume, and degree of cleanliness o...
Toimittaja: VWR Collection
Kuvaus: B'mat decontamination flooring uses a patented polymer to attract and retain over 98% of foot- and wheel-borne particles. Particles are naturally attr...
Tuoteluettelon nro: (590-0346)
Toimittaja: Bohlender
Kuvaus: Mat made of static dissipative material with permanently mounted grounding cable including clip for earthing connection.
UOM: 1 * 1 Items
Toimittaja: 3M
Kuvaus: The Nomad™ Terra Heavy Traffic Entrance Matting 8100 is unbacked scraper matting suitable for use indoors, outdoors, or in recessed wells with high tr...
Toimittaja: 3M
Kuvaus: The Nomad™ Aqua Textile Drop Down Mat 85 is a high-performance entrance mat with a phthalate-free, slip-resistant PVC backing for high traffic applica...
Toimittaja: 3M
Kuvaus: The Nomad™ Aqua Textile Drop Down Mat 65 is a high-performance entrance mat designed for medium traffic applications.
Toimittaja: 3M
Kuvaus: The Nomad™ Aqua Textile Drop Down Mat 45 is a high-performance entrance mat backed with slip-resistant PVC for low traffic applications.
Tuoteluettelon nro: (NEWPFLME175)
Toimittaja: NEW PIG
Kuvaus: Clean and sanitise shoes and prevent contamination from entering controlled areas with this disinfecting footbath.
UOM: 1 * 1 Items
Toimittaja: 3M
Kuvaus: The Nomad™ Terra Medium Traffic Entrance Matting 6050 is backed vinyl matting suitable for use in indoor areas with medium traffic volumes.
Toimittaja: 3M
Kuvaus: 3M™ Nomad™ terra heavy traffic entrance matting 8250 is a coiled vinyl scraper mat suitable for use indoors, outdoors, or in recessed wells with high ...
New Product
Toimittaja: DYCEM
Kuvaus: These floating floor mats are high performance, antimicrobial loose-laid mats offering many benefits without the need for permanent installation. A fl...
Tuoteluettelon nro: (DYCECZ01/WZP/2GRAN)
Toimittaja: DYCEM
Kuvaus: These mats are specially designed to remove contamination from areas subject to heavy wheeled traffic. Ideal for use at entrances and exits, corridors...
UOM: 1 * 2 m2
Toimittaja: DYCEM
Kuvaus: Specially designed mat to remove contamination from areas subject to heavy wheeled traffic. It effectively attracts, collects and retains contaminatio...
Tuoteluettelon nro: (DYCECZ06/BM60045GR)
Toimittaja: DYCEM
Kuvaus: This non-slip mat prevents the movement and loss of small components used in laboratories to stabilise fragile equipment. Approved for use in all clas...
UOM: 1 * 1 Items
Toimittaja: asecos
Kuvaus: Sealing mats made from a dense and flexible material prevent fast and efficient penetration of water-polluting liquids into the ground or drains.
Toimittaja: DYCEM
Kuvaus: CleanZone is a high performance contamination control flooring suitable for light wheeled traffic and heavier pedestrian flows. It is washable and a mos...
Tiedustele hintaa asiakaspalvelusta.
Tämän nimikkeen varasto on rajallinen, mutta sitä saattaa olla sinua lähellä sijaitsevassa varastossa. Varmista, että olet kirjautunut sisään, jotta voit nähdä varaston tilan. Jos call näkyy yhä ja tarvitset apua, soita numeroon (09) 8045 5300.
Tämän nimikkeen varasto on rajallinen, mutta sitä saattaa olla sinua lähellä sijaitsevassa varastossa. Varmista, että olet kirjautunut sisään, jotta voit nähdä varaston tilan. Jos call näkyy yhä ja tarvitset apua, soita numeroon (09) 8045 5300.
Tämä tuote on luvanvarainen.
Tarvitessamme lisätietoja otamme teihin yhteyttä sähköpostitse.
Alkoholi ja käyttöluvat: Asiakkaalla on oltava kyseisessä toimipaikassa voimassa oleva lupaviranomaisen (Valvira) myöntämä alkoholin ostolupa tilaamilleen tuotteille.
Tämä tuote on luvanvarainen.
Tarvitessamme lisätietoja otamme teihin yhteyttä sähköpostitse.
Alkoholi ja käyttöluvat: Asiakkaalla on oltava kyseisessä toimipaikassa voimassa oleva lupaviranomaisen (Valvira) myöntämä alkoholin ostolupa tilaamilleen tuotteille.
Organisaatiosi on estänyt tämän tuotteen hankkimisen. Saat lisätietoja omalta hankintaosastoltasi.
Tuotetta ei ole enää saatavana. Korvaava tuote näkyy alla.
Tällä symbolilla merkityt tuotteet poistetaan - varasto myydään loppuun. Mahdollisia vaihtoehtoja voi etsiä yllä näkyvillä VWR tuotenumeroilla. Jos tarvitset lisäapua, ota yhteyttä info.fi@vwr.com
1 - 16 of 17
no targeter for Bottom