6-Metoksi-1-tetraloni

Toimittaja: Molekula

MOLE27116667-100GEA 181 EUR
MOLE27116667-100G
6-Metoksi-1-tetraloni
6-Metoksi-1-tetraloni
Kaava: C₁₁H₁₂O₂
MW: 176,22 g/mol
MDL numero: MFCD00001695
CAS-numero: 1078-19-9
EINECS: 214-078-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR