Multi-purpose stand with weighted base and flexible arm. One end is designed to accept the electrode holder (sold separately) while the other has a 1,3 cm diameter stainless steel pin for attachment to the weighted base or a suitable clamp.
- Flexible arm with molded acetal copolymer links has ball-and-socket joints
- 20 individual links provide maximum range of motion
- Octagonal-shaped wells (4 total) are centered along each edge of the base
- Electrode holder snaps onto the end of the flexible gooseneck arm affording three stepped wells for holding an electrode, a reference electrode and an ATC probe
- Center well of the holder has an 1,8 cm int.Ø, side wells each have a 1,7 cm int.Ø; through holes are all 1,3 cm int.Ø
Flexible arm measures 46×2,5 cm Ø.
Sturdy base measures 23×16 cm and weighs 1,5 kg.
Toimitustiedot: Supplied with three reducing bushings for smaller diameters and five nylon cable ties for attaching wires to the arm.