Electrode stand, SP Bel-Art
Toimittaja: Bel-Art Products, a part of SP
212-6623EA 105 EUR
212-6623 BELAF183150000
Electrode stand, SP Bel-Art
Elektroditeline
Multi-purpose stand with weighted base and flexible arm. One end is designed to accept the electrode holder (sold separately) while the other has a 1,3 cm diameter stainless steel pin for attachment to the weighted base or a suitable clamp.
  • Flexible arm with molded acetal copolymer links has ball-and-socket joints
  • 20 individual links provide maximum range of motion
  • Octagonal-shaped wells (4 total) are centered along each edge of the base
  • Electrode holder snaps onto the end of the flexible gooseneck arm affording three stepped wells for holding an electrode, a reference electrode and an ATC probe
  • Center well of the holder has an 1,8 cm int.Ø, side wells each have a 1,7 cm int.Ø; through holes are all 1,3 cm int.Ø
Flexible arm measures 46×2,5 cm Ø.
Sturdy base measures 23×16 cm and weighs 1,5 kg.
Toimitustiedot: Supplied with three reducing bushings for smaller diameters and five nylon cable ties for attaching wires to the arm.