Fraction collectors, CF-2
Toimittaja: Spectrum
SPEC124846EA 6200 EUR
SPEC124846
Fraction collectors, CF-2
Fraktionkerääjät
The CF-2 fraction collector is a very durable general-purpose fraction collector that meets both preparative LC and open column LC needs.
  • Features rugged drive and rugged matallic casework
  • Cold room compatible
  • CF-2 is sold complete and ready-to-use
The CF-2 Fraction Collector is designed with a stationary drop head where the tubes move under the drop former insuring every drop is collected. It includes a powerful peak detector that used to separate the low-level 'in-between' material from the peaks.
Toimitustiedot: Supplied with tube holders for 12 or 13 mm diameter tubes and/or a drop counter and tubing fittings.