Alusvaate, pusero, 789 OF
Toimittaja: Fristads Kansas Group
Match
KWIN100419940LEA 40.6 EUR
KWIN100419940L KWIN1004199402XL KWIN100419940XS KWIN100419940M KWIN1004199403XL KWIN100419940S KWIN100419940XL
Alusvaate, pusero, 789 OF
Työvaatteet
Moisture-wicking and higly breathble shirt with long sleeves made of 100% polyester.
  • Quick drying
  • Polo-neck with zip
  • Grey contrast seams
  • Machine washable up to 60 °C
Weight: 160 g/m²
Sertifikaatit: OEKO-TEX® certified.