Poolopaita, ESD, diamond knit, XF83
Toimittaja: Fristads Kansas Group
Fristads®
113-05403EA 0 EUR
113-05403 113-05404 113-05400 113-05401 113-05402
Poolopaita, ESD, diamond knit, XF83
Työvaatteet
ESD shirt manufactured from a knitted blend (83% cotton, 14% polyester and 3% conductive fibre), which makes it comfortable to wear. The special knitting technique produces good conductivity although the content of conductive fibre is only 3%. The garments are tested and certified under IEC 61340.
  • Short-sleeved
  • Polo collar
  • Three snap fasteners
Weight: 160 g/m²