Fixation plasters, Askina® Film
Toimittaja: B. Braun
Askina® Film
BRAU9081500EA 0 EUR
BRAU9081500
Fixation plasters, Askina® Film
Ensiaputarvikkeet Haavasidokset, laastarit
Perforated foil plaster for the fixation of all types of wound dressings. Also suitable for the fixation of catheters, tubes and puncture sets.
  • Mouldable, perforated foil material
  • Air and water vapor permeable
  • Transparent