D-(-)-Amethopterin hydrate

Toimittaja: Molekula

MOLEM36015135EA 139 EUR
MOLEM36015135
D-(-)-Amethopterin hydrate
D-(-)-Amethopterin hydrate
Kaava: C₂₀H₂₂N₈O₅·H₂O
CAS-numero: 331717-43-2
EINECS: 257-482-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR