3,5,5-Trimethyloxazolidine-2,4-dione (Trimethadione)

EDQMT2100000EA 282 EUR
EDQMT2100000
3,5,5-Trimethyloxazolidine-2,4-dione (Trimethadione)
3,5,5-Trimethyloxazolidine-2,4-dione (trimethadione)
Kaava: C₆H₉NO₃
MW: 143,1406 g/mol
MDL numero: MFCD00047084
CAS-numero: 127-48-0
EINECS: 204-845-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR