L(-)-Carnitine

EDQML0399900EA 282 EUR
EDQML0399900
L(-)-Carnitine
L(-)-Carnitine
Kaava: C₇H₁₅NO₃
MW: 161,20 g/mol
MDL numero: MFCD00038747
CAS-numero: 541-15-1
EINECS: 208-768-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR