Ice pans and buckets, multi-purpose
BCISBCS-115-25GREA 103.8 EUR
BCISBCS-115-25GR BCISBCS-211PK BCISBCS-115GR BCISBCS-115-25R BCISBCS-115 BCISBCS-112 BCISBCS-113 BCISBCS-113OR BCISBCS-115-25OR BCISBCS-115OR BCISBCS-111GR BCISBCS-115-25PL BCISBCS-115-25PK BCISBCS-211GR BCISBCS-115PK BCISBCS-115-25G BCISBCS-115PL BCISBCS-111OR BCISBCS-111PK
Ice pans and buckets, multi-purpose
Ethylene vinyl acetate (EVA) foam
  • Solid, one piece moulded, unbreakable
  • Resistant to highly acidic and basic solutions, salts, alcohol, acetone, chloroform, phenol, etc.
  • Insulative properties and resistant to temperatures from −196 to 93 °C
  • Available in choice of colours and sizes
Ideal for use with ice, dry ice, liquid nitrogen, saline solution and acetone.