Cover spectacles, Horizon
Toimittaja: Honeywell Safety
Horizon Blue
DALL3024130-STDEA 41.8 EUR
DALL3024130-STD
Cover spectacles, Horizon
Suojalasit
The highly technical cover glasses (OTG) are made from a polyamide frame and moulded sideshields.
  • Easily adjustable temples
  • Integrated superior and sub-orbital protection
  • Can be equipped with four types of flip-up windows depending on the application
Frame designed to accept prescription lenses.
Weight: 34 g.