2,3,5,6-Tetrafluoritiofenoli ≥95.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAT1044-5GEA 72.6 EUR
TCIAT1044-5G
2,3,5,6-Tetrafluoritiofenoli ≥95.0% (by GC)
2,3,5,6-Tetrafluoritiofenoli
Kaava: C₆H₂F₄S
MW: 182,14 g/mol
Kiehumispiste: 147°C
Leimahduspiste: 48°C
MDL numero: MFCD00004829
CAS-numero: 769-40-4
EINECS: 212-210-1
UN: 1993
ADR: 3,III
https://fi.vwr.com/stibo/web/std.lang.all/51/17/28995117.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR