Metyleenisininen ≥70% (W)

Toimittaja: TCI EUROPE
Vaara

Synonyms: C.I. 52015Basic Blue 9

TCIAM0501-25GEA 29.7 EUR
TCIAM0501-25G
Metyleenisininen ≥70% (W)
Metyleenisininen
Kaava: C₁₆H₁₈ClN₃S·xH₂O
MW: 319,86 g/mol (anhy)
MDL numero: MFCD00012111
CAS-numero: 122965-43-9
EINECS: 200-515-2
https://fi.vwr.com/stibo/web/std.lang.all/39/62/28973962.jpg

Order Now

Testitulosten erittely

Appearance Green to Dark green powder to crystal
Purity (Gravimetric Method) min. 70.0 % (calcd.on dried substance)
Lambda max. 660.0 to 665.0 nm (EtOH (50%))
Absorbance (E1%1cm) min. 2000 (EtOH (50%), 660.0 to 665.0 nm)
Drying loss max. 20.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR