Cyclohexyl propionate ≥98.0% (by GC)
Toimittaja: TCI EUROPE
TCIAP0504-25MLEA
30.8
EUR
TCIAP0504-25ML
Cyclohexyl propionate ≥98.0% (by GC)
Cyclohexyl propionate
Kaava:
C₉H₁₆O₂ MW: 156,22 g/mol Kiehumispiste: 193°C Sulamispiste: –33 °C Leimahduspiste: 76°C |
MDL numero:
MFCD00046352 CAS-numero: 6222-35-1 EINECS: 228-303-5 |
Testitulosten erittely
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...