5-Hydroksi-isoftaalihappo ≥97.0% (by titrimetric analysis)
Toimittaja: TCI EUROPE
Varoitus
TCIAH0794-500GEA 504 EUR
TCIAH0794-500G TCIAH0794-25G
5-Hydroksi-isoftaalihappo ≥97.0% (by titrimetric analysis)
5-Hydroksi-isoftaalihappo
Kaava: C₈H₆O₅
MW: 182,13 g/mol
Sulamispiste: 298…301 °C
MDL numero: MFCD00002515
CAS-numero: 618-83-7
EINECS: 210-565-7
https://fi.vwr.com/stibo/web/std.lang.all/08/25/28970825.jpg
Order Now
Testitulosten erittely
Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Solubility in Methanol almost transparency