ω,ω-Diklooriasetofenoni ≥97.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAD1122-25GEA 174 EUR
TCIAD1122-25G
ω,ω-Diklooriasetofenoni ≥97.0% (by GC)
ω,ω-Diklooriasetofenoni
Kaava: (C₆H₅)C(O)CHCl₂
MW: 189,04 g/mol
Sulamispiste: 23…24 °C
MDL numero: MFCD00000844
CAS-numero: 2648-61-5
https://fi.vwr.com/stibo/web/std.lang.all/63/56/28976356.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR