1-Bromo-2-naphthol ≥97.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAB0620-25GEA 115 EUR
TCIAB0620-25G
1-Bromo-2-naphthol ≥97.0% (by GC)
1-Bromo-2-naphthol
Kaava: C₁₀H₇BrO
MW: 223,07 g/mol
Sulamispiste: 79…81 °C
MDL numero: MFCD00003869
CAS-numero: 573-97-7
EINECS: 209-363-1
https://fi.vwr.com/stibo/web/std.lang.all/25/51/28932551.jpg

Order Now

Testitulosten erittely

Appearance White to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 78.0 to 84.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR