1-naftolibentseiini
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAN0028-25GEA
220
EUR
TCIAN0028-25G
1-naftolibentseiini
1-naftolibentseiini
Kaava:
C₂₇H₁₈O₂ MW: 374,44 g/mol Sulamispiste: 243…245 °C |
MDL numero:
MFCD00078492 CAS-numero: 145-50-6 EINECS: 205-656-3 |
Testitulosten erittely
Appearance | Orange to Amber to Dark red powder to crystal |
Absorbance | min. 0.600 (10 mg/L, pH 12.0, 648.0 to 652.0 nm) |
Sensitiveness | Turn Reddish yellow in Acetic Acid and turn green by adding Perchloric Acid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...