8-Anilino-1-naphthalenesulphonic acid ≥95.0% (by HPLC, titration analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAA0472-25GEA 62.7 EUR
TCIAA0472-25G
8-Anilino-1-naphthalenesulphonic acid ≥95.0% (by HPLC, titration analysis)
8-Anilino-1-naphthalenesulphonic acid
Kaava: C₁₆H₁₃NO₃S
MW: 299,35 g/mol
MDL numero: MFCD00003998
CAS-numero: 82-76-8
EINECS: 201-438-7
https://fi.vwr.com/stibo/web/std.lang.all/79/03/28927903.jpg

Order Now

Testitulosten erittely

Appearance Light gray to Brown to Black powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR