Methyl green (Basic Blue 20)

Toimittaja: TCI EUROPE

TCIAM0498-25GEA 145 EUR
TCIAM0498-25G
Methyl green (Basic Blue 20)
Methyl green (Basic Blue 20)
Kaava: C₂₆H₃₃Cl₂N₃
MW: 458,47 g/mol
MDL numero: MFCD00011879
CAS-numero: 82-94-0
EINECS: 201-447-6
https://fi.vwr.com/stibo/web/std.lang.all/39/61/28973961.jpg

Order Now

Testitulosten erittely

Appearance Dark green to Dark blue to Black powder to crystal
Lambda max. 633.0 to 637.0 nm (EtOH (50%))
Absorbance (E1%1cm) min. 350 (EtOH (50%), 633.0 to 637.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR