1-Naphthonitrile ≥95.0% (by GC)
Toimittaja: TCI EUROPE
|
Vaara
|
TCIAN0038-25GEA
66
EUR
TCIAN0038-25G
TCIAN0038-250G
1-Naphthonitrile ≥95.0% (by GC)
1-Naphthonitrile
Kaava:
C₁₁H₇N MW: 153,18 Kiehumispiste: 299°C |
MDL numero:
MFCD00003866 CAS-numero: 86-53-3 EINECS: 201-679-8 UN: 3439 ADR: 6.1,III |
![]() |
Testitulosten erittely
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 95.0 % |
Freezing point | 30.0 to 36.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...