(S)-(+)-Nipecotic acid

Toimittaja: Molekula

MOLEM34397408EA 127 EUR
MOLEM34397408
(S)-(+)-Nipecotic acid
(S)-(+)-Nipecotic acid
Kaava: C₆H₁₁NO₂
MW: 129,16
MDL numero: MFCD01630807
CAS-numero: 59045-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR