3-Bromo-5-(pentafluorothio)aniline ≥97%

Varoitus

H34456.MDEA 104 EUR
H34456.MD H34456.03
3-Bromo-5-(pentafluorothio)aniline ≥97%
3-Bromo-5-(pentafluorothio)aniline
Kaava: C₆H₅BrF₅NS
MW: 298,07
Storage Temperature: Huoneenlämpö
MDL numero: MFCD16652416
CAS-numero: 1240257-95-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR