2-Methyl-5-(pentafluorothio)benzoic acid ≥97%

Varoitus

H33071.03EA 503 EUR
H33071.03 H33071.MD
2-Methyl-5-(pentafluorothio)benzoic acid ≥97%
2-Methyl-5-(pentafluorothio)benzoic acid
Kaava: C₈H₇F₅O₂S
MW: 262,19 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD16652477
CAS-numero: 623943-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR