4-(4-Nitrophenylazo)-1-naphthol

Toimittaja: Molekula

MOLE32826669-25GEA 0 EUR
MOLE32826669-25G
4-(4-Nitrophenylazo)-1-naphthol
4-(4-Nitrophenylazo)-1-naphthol
Kaava: C₁₆H₁₁N₃O₃
MW: 293,28 g/mol
MDL numero: MFCD00003975
CAS-numero: 5290-62-0
EINECS: 226-129-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR