2,5-Diphenyl-1,3,4-oxadiazole

Toimittaja: Molekula

MOLE35371792-5GEA 0 EUR
MOLE35371792-5G
2,5-Diphenyl-1,3,4-oxadiazole
2,5-Diphenyl-1,3,4-oxadiazole
Kaava: C₁₄H₁₀N₂O
MDL numero: MFCD00003102
CAS-numero: 725-12-2
EINECS: 211-968-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR