Automatic burettes, Pellet pattern, Schellbach
Toimittaja: Hecht
HECH48415015EA 325 EUR
HECH48415015 HECH48415050 HECH48415022
Automatic burettes, Pellet pattern, Schellbach
Byretit
Automatic burettes, conformity certified DURAN® borosilicate glass 3.3, with stopcock between burette and bottle.
  • Class AS, with Schellbach stripe, fused blue graduation
  • Delivery time 35 to 45 seconds
  • With reservoir bottle 2000 ml with standard ground joint NS 29/32
  • Rubber bulb provided
Sertifikaatit: Available with individual certificate of performance, USP batch certificate or DAkkS calibration certificate.