Automatic burettes, Pellet pattern, without intermediate stopcock
Toimittaja: Hecht
HECH48462025EA 283 EUR
HECH48462025 HECH48462050
Automatic burettes, Pellet pattern, without intermediate stopcock
Byretit
Automatic burettes, made of DURAN® borosilicate glass.
  • Fused amber graduation
  • Acid and alkali resistant
  • Stopcock with interchangeable safety glass keys
With reservoir bottle 2000 ml with standard ground jont and rubber bulb.