Sperm measuring cylinders, with funnel top
Toimittaja: Hecht
HECH42220010EA 14.1 EUR
HECH42220010
Sperm measuring cylinders, with funnel top
Glass, clear.
  • With funnel top
  • Approximate scale
  • Fused amber graduation